missing translation for 'onlineSavingsMsg'
Learn More

CD31 (PECAM-1) Mouse anti-Human, Super Bright 600, Clone: WM-59 (WM59), eBioscience™

Mouse Monoclonal Antibody

141.00€ - 301.00€

Especificaciones

Clasificación Monoclonal
Monoclonal o policlonal Monoclonal
Primario o secundario Primary
Antígeno CD31 (PECAM-1)
Conjugado Super Bright 600
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad. Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad. Precio Cantidad y disponibilidad  
15933494 Invitrogen
63-0319-42
100 Tests 301.00€
paquete de 100 Ver productos alternativos
Ahorre hasta 
»
Fecha de envío estimada
del proveedor 05-06-2019
Añadir a la cesta
 
15923494 Invitrogen
63-0319-41
25 Tests 141.00€
paquete de 25 Ver productos alternativos
Ahorre hasta 
»
Fecha de envío estimada
del proveedor 05-06-2019
Añadir a la cesta
 
Descripción

Descripción

Please refer to the datasheet for Super Bright Staining Buffer for more information. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 600 nm; Laser: Violet Laser CD31 (platelet endothelial cell adhesion molecule-1, PECAM-1) is an inhibitory coreceptor involved in regulation of T cell and B cell signaling by a dual immunoreceptor tyrosine-based inhibitory motif (ITIM) that upon associated kinases-mediated phosphorylation provide docking sites for protein-tyrosine phosphatases. CD31 is expressed ubiquitously within the vascular compartment and is located mainly at junctions between adjacent cells. N-terminal Ig-like domain of CD31 is responsible for its homophilic binding, which plays an important role in cell-cell interactions. CD31 is a multifunctional molecule with diverse roles in modulation of integrin-mediated cell adhesion, transendothelial migration, angiogenesis, apoptosis, negative regulation of immunoreceptor signaling, autoimmunity, macrophage phagocytosis, IgE-mediated anaphylaxis and thrombosis. It is one of key regulatory molecules in vascular system.
Especificaciones

Especificaciones

Monoclonal
Primary
Super Bright 600
Monoclonal
CD31 (PECAM-1)