Learn More
CD31 (PECAM-1) Mouse anti-Human, Super Bright 436, Clone: WM-59 (WM59), eBioscience™
Mouse Monoclonal Antibody
Marca:  Invitrogen 62-0319-41
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 15933374
  • 126.00€ / paquete de 25
Fecha de envío estimada
del proveedor 23-10-2019
Añadir a la cesta
Descripción
Descripción
Please refer to the datasheet for Super Bright Staining Buffer for more information. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company. CD31 (platelet endothelial cell adhesion molecule-1, PECAM-1) is an inhibitory coreceptor involved in regulation of T cell and B cell signaling by a dual immunoreceptor tyrosine-based inhibitory motif (ITIM) that upon associated kinases-mediated phosphorylation provide docking sites for protein-tyrosine phosphatases. CD31 is expressed ubiquitously within the vascular compartment and is located mainly at junctions between adjacent cells. N-terminal Ig-like domain of CD31 is responsible for its homophilic binding, which plays an important role in cell-cell interactions. CD31 is a multifunctional molecule with diverse roles in modulation of integrin-mediated cell adhesion, transendothelial migration, angiogenesis, apoptosis, negative regulation of immunoreceptor signaling, autoimmunity, macrophage phagocytosis, IgE-mediated anaphylaxis and thrombosis. It is one of key regulatory molecules in vascular system.
Especificaciones
Especificaciones
CD31 (PECAM-1)
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
P16284
CD31; CD31/EndoCAM; endoCAM; GPIIA′; PECA1; PECAM-1; PECAM1
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
WM-59 (WM59)
Super Bright 436
PECAM1
Liquid
PECAM1
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
5175