Learn More
CD31 Mouse anti-Human, Clone: 2A1E2, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66065-1-IG-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
PECAM-1 (Platelet endothelial cell adhesion molecule), also known as cluster of differentiation 31 (CD31) is a protein that in human is encoded by the PECAM1 gene found on chromosome 17. PECAM1 is a member of the immunoglobulin (Ig) superfamily that is expressed on the surface of circulating platelets, monocytes, neutrophils, and particular T-cell subsets. Using a PCR-based analysis of somatic cell hybrids, Gumina et al. (1996) mapped PECAM1 to chromosome 17 in the region 17q23-qter. Several adhesion molecules expressed on platelets and endothelium also localized to 17q. Xie and Muller (1996) mapped the Pecam1 gene to mouse chromosome 6, region F3-G1, by fluorescence in situ hybridization. PECAM-1 is found on the surface of platelets, monocytes, neutrophils, and some types of T-cells, and makes up a large portion of endothelial cell intercellular junctions, and PECAM-1 plays a key role in removing aged neutrophils from the body.Especificaciones
CD31 | |
Monoclonal | |
0.91 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P16284 | |
PECAM1 | |
CD31 Fusion Protein Ag1787 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
2A1E2 | |
Unconjugated | |
PECAM1 | |
CD31, EndoCAM, GPIIA, PECA1, PECAM 1, PECAM1, VEC marker | |
Mouse | |
Protein A | |
RUO | |
5175 | |
-20°C | |
Liquid |