missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse, FITC, Clone: Hit3a, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio QAB7-F-100Tests
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CD3 | |
Monoclonal | |
5μL/Test | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
CD3E | |
Protein A/G Purified | |
RUO | |
916 | |
IgG2a κ |
Flow Cytometry | |
Hit3a | |
FITC | |
NP_000724 | |
Mouse | |
100 Tests | |
Primary | |
Store at 2-4°C. |