missing translation for 'onlineSavingsMsg'
Learn More

CD3 delta Antibody (138), Alexa Fluor™ 488, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-89730AF488

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213279

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

CD3 delta Monoclonal specifically detects CD3 delta in Human samples. It is validated for Western Blot.
Especificaciones

Especificaciones

CD3 delta
Monoclonal
Alexa Fluor 488
CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, T3DOKT3, delta chain, T-cell receptor T3 delta chain, T-cell surface glycoprotein CD3 delta chain
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD3 delta (NP_000723.1; Met1-Ala105).
0.1 mL
Adaptive Immunity, Apoptosis, Diabetes Research, Immunology, Innate Immunity, Signal Transduction
915
Store at 4°C in the dark.
IgG
Western Blot
138
50 mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales