missing translation for 'onlineSavingsMsg'
Learn More

CD28 Mouse anti-Human, Super Bright 436, Clone: CD28.2, eBioscience™

Mouse Monoclonal Antibody

Marca:  Invitrogen 62-0289-41

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15993364

  • 145.00€ / paquete de 25
Envío estimado: 02-02-2022

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Description: The CD28.2 monoclonal antibody reacts with the human CD28 molecule, a 44 kDa homodimer expressed by thymocytes, mature T cells and plasma cells. CD28 is a ligand for CD80 (B7-1) and CD86 (B7-2) and is a potent co-stimulator of T cells. Signaling through CD28 augments IL-2 and IL-2 receptor expression as well as cytotoxicity of CD3-activated T cells. Applications Reported: This CD28.2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This CD28.2 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No. SB-4400) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser

Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company. CD28 antigen is a 44 kDa disulfide linked homodimeric T cell specific surface glycoprotein. CD28 is a cell adhesion molecule of the immunoglobulin superfamily which is constitutively expressed on most peripheral blood T lymphocytes. Moreover, CD28 is the critical T cell costimulatory receptor that provides the cell the important second activation signal by binding CD80 and CD86 which are expressed by antigen presenting cells. In addition to its co-stimulation role, CD28 functions by preventing T cells from entering an anergic-hyporesponsive state or from undergoing premature apoptotic cell death. In murine peripheral lymphoid organs and in the blood, all CD4+ and CD8+ T cells express CD28. In the thymus, CD28 expression is highest on immature CD3-, CD8+ and CD4+8+ cells, and on CD4-8- cells that express alpha/beta and gamma/delta TCR. The level of CD28 on mature CD4+ and CD8+ alpha/beta TCR+ thymocytes is two- to fourfold lower than on the immature cells. Diseases associated with CD28 dysfunction include mycosis fungiodes and Sezary’s Disease.
Especificaciones

Especificaciones

CD28
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
P10747
CD28; CD28 antigen; MGC138290; T-cell-specific surface glycoprotein CD28; TP44
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
CD28.2
Super Bright 436
CD28
Liquid
CD28
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
940
Videos
SDS
Documentos
Certificados
Ofertas especiales

Ofertas especiales