missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD279 / PD-1 Armenian Hamster, PE, Clone: J43.1, enQuireBio™
Armenian Hamster Monoclonal Antibody
Marca: EnquireBio QAB98-PE-25ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CD279 / PD-1 | |
Monoclonal | |
0.2mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Pdcd1 | |
Protein A/G Purified | |
RUO | |
18566 | |
IgG |
Flow Cytometry | |
J43.1 | |
PE | |
NP_032824 | |
Armenian Hamster | |
25 μg | |
Primary | |
Store at 2-4°C. |