missing translation for 'onlineSavingsMsg'
Learn More

CD274 (PD-L1, B7-H1) Rat anti-Mouse, Super Bright 645, Clone: MIH5, eBioscience™

Rat Monoclonal Antibody

Marca:  Invitrogen 64-5982-82

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15933674

  • 316.00€ / 100 microgramos


Fecha estimada de envío
17-07-2020
Añadir a la cesta

Descripción

Descripción

SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333) for up to 3 days in the dark at 4 degrees C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 645 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company. Programmed death receptor ligand 1 (PD-L1, also called B7-H1) is a recently described B7 family member. To date, one specific receptor has been identified that can be ligated by PD-L1. This receptor, programmed death receptor 1 (PD-1), has been shown to negatively regulate T-cell receptor (TCR) signaling. Upon ligating its receptor, PD-L1 has been reported to decrease TCR-mediated proliferation and cytokine production. PD-L1 expression was found to be abundant on many murine and human cancers and could be further up-regulated upon IFN-gamma stimulation. Thus, PD-L1 might play an important role in tumor immune evasion.
Especificaciones

Especificaciones

CD274 (PD-L1, B7-H1)
Monoclonal
0.2 mg/mL
PBS with BSA and 0.09% sodium azide; pH 7.2
Q9EP73
B7 homolog 1; B7-H; B7-H1; B7H1; CD274; CD274 antigen; PD-L1; PDCD1 ligand 1; PDCD1L1; PDCD1LG1; PDL1; Programmed cell death 1 ligand 1; programmed death ligand 1
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
MIH5
Super Bright 645
Cd274
Liquid
Cd274
IgG2a, lambda
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
60533
Videos
Documentos