missing translation for 'onlineSavingsMsg'
Learn More

CD25 Mouse anti-Canine, Super Bright 600, Clone: P4A10, eBioscience™

Mouse Monoclonal Antibody

Marca:  Invitrogen 63-0250-41

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15933484

  • 141.00€ / paquete de 25


Fecha de envío estimada
del proveedor 02-10-2019
Añadir a la cesta

Descripción

Descripción

When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No. SB-4400) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 600 nm; Laser: Violet Laser CD25 (IL2 receptor alpha chain/IL2RA) is a cytokine that plays a role in the proliferation of T and B lymphocytes. The receptor of this cytokine (IL2RA) is a heterotrimeric protein complex with a gamma chain also shared by interleukin 4 (IL4) and interleukin 7 (IL7). IL2RA, IL2R beta chain (IL2RB), and the IL2R gamma chain (IL2RG), constitute the high-affinity IL2 receptor. Homodimeric IL2RA chains result in low-affinity receptor, while homodimeric IL2RB chains produce a medium-affinity receptor. The expression of IL2 in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. IL2 is primarily produced by mature T cells. IL2 plays an important role as a growth factor, differentiation factor, and regulator of cell death. IL-2 stimulates the proliferation of B cells, augments natural killer cell activity, and inhibits granulocyte macrophage colony formation. The targeted disruption of a similar gene in mice leads to ulcerative colitis-like disease, which suggests a role in the immune response to antigenic stimuli. Mutations in this gene are associated with interleukin 2 receptor alpha deficiency.
Especificaciones

Especificaciones

CD25
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
O62802
IL2RA
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
403870
Flow Cytometry
P4A10
Super Bright 600
IL2RA
Liquid
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Canine
Videos
Documentos