missing translation for 'onlineSavingsMsg'
Learn More

CD24, Super Bright 600, Clone: eBioSN3 (SN3 A5-2H10), Monoclonal, eBioscience™

Mouse Monoclonal Antibody

Marca:  Invitrogen 63-0247-41

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15913484

  • 150.00€ / paquete de 25


Fecha de envío estimada
del proveedor 03-05-2019
Añadir a la cesta

Descripción

Descripción

Please refer to the datasheet for Super Bright Staining Buffer for more information. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 600 nm; Laser: Violet Laser CD24 is a two-chain glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed at multiple stages of B-cell development, beginning with the bone marrow pro-B-cell compartment and continuing through mature, surface Ig positive B-cells. Plasma cell expression is very low or negative. CD24 is also expressed on the majority of B-lineage acute lymphoblastic leukemias, B-cell CCLs and B-cell non-Hodgkin′s lymphomas. CD24 may play a role in regulation of B-cell proliferation and maturation, and control of autoimmunity.
Especificación

Especificación

CD24
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
P25063
CD24
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
100133941
Flow Cytometry
eBioSN3 (SN3 A5-2H10)
Super Bright 600
CD24
Liquid
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Videos