missing translation for 'onlineSavingsMsg'
Learn More

CD24 Rat anti-Mouse, Super Bright 436, Clone: M1/69, eBioscience™

Rat Monoclonal Antibody

Marca:  Invitrogen 62-0242-80

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15913364

  • 109.00€ / 25 microgramos


Fecha de envío estimada
del proveedor 03-05-2019
Añadir a la cesta

Descripción

Descripción

When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (cat. SB-4400) to minimize any non-specific polymer interactions. Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser CD24 is a two-chain glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed at multiple stages of B-cell development, beginning with the bone marrow pro-B-cell compartment and continuing through mature, surface Ig positive B-cells. Plasma cell expression is very low or negative. CD24 is also expressed on the majority of B-lineage acute lymphoblastic leukemias, B-cell CCLs and B-cell non-Hodgkin′s lymphomas. CD24 may play a role in regulation of B-cell proliferation and maturation, and control of autoimmunity.
Especificación

Especificación

CD24
Monoclonal
0.2 mg/mL
PBS with BSA and 0.09% sodium azide; pH 7.2
P24807
CD24 antigen (small cell lung carcinoma cluster 4 antigen); CD24A; FLJ22950; FLJ43543; MGC75043
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
M1/69
Super Bright 436
Cd24a
Liquid
Cd24a
IgG2b, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
12484
Videos