missing translation for 'onlineSavingsMsg'
Learn More

CD23 Mouse anti-Human, Mouse, Clone: 3H6B8, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 60208-1-IG-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16827215

  • 140.00€ / 20 microlitros
Fecha estimada de envío: 02-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

CD23 is a 45 kDa glycoprotein which is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. The CD23 molecule is identical to the low affinity IgE receptor found on B cells. Expression of CD23 has been detected in neoplastic cells from cases of B cell chronic lymphocyctic leukaemia and some cases of centroblastic/centrocytic lymphoma. CD23 is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. Functionally, CD23 is involved in B cell growth and differentiation, and IgE production. Further, CD23 has a soluble form that is a potent mitogenic factor. Diseases associated with CD23 dysfunction include chronic conjunctivitis and chronic lymphocytic leukemia.
Especificaciones

Especificaciones

CD23
Monoclonal
0.51 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P06734, P20693
FCER2
CD23 Fusion Protein Ag0425
20 μL
Primary
Human, Mouse
Antibody
IgG1
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin)
3H6B8
Unconjugated
FCER2
BLAST 2, CD23, CD23A, CLEC4J, Fc epsilon RII, FCE2, FCER2, IGEBF, Lymphocyte IgE receptor
Mouse
Protein G
RUO
14128, 2208
-20°C
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales