missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD23 Mouse anti-Human, Mouse, Clone: 3H6B8, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60208-1-IG-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
CD23 is a 45 kDa glycoprotein which is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. The CD23 molecule is identical to the low affinity IgE receptor found on B cells. Expression of CD23 has been detected in neoplastic cells from cases of B cell chronic lymphocyctic leukaemia and some cases of centroblastic/centrocytic lymphoma. CD23 is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. Functionally, CD23 is involved in B cell growth and differentiation, and IgE production. Further, CD23 has a soluble form that is a potent mitogenic factor. Diseases associated with CD23 dysfunction include chronic conjunctivitis and chronic lymphocytic leukemia.Especificaciones
CD23 | |
Monoclonal | |
0.51 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P06734, P20693 | |
FCER2 | |
CD23 Fusion Protein Ag0425 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
3H6B8 | |
Unconjugated | |
FCER2 | |
BLAST 2, CD23, CD23A, CLEC4J, Fc epsilon RII, FCE2, FCER2, IGEBF, Lymphocyte IgE receptor | |
Mouse | |
Protein G | |
RUO | |
14128, 2208 | |
-20°C | |
Liquid |