missing translation for 'onlineSavingsMsg'
Learn More

CD229/SLAMF3/Lymphocyte Antigen 9 Antibody (002), Alexa Fluor™ 647, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Bio-Techne NBP2-90617AF647

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215247

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

CD229/SLAMF3/Lymphocyte Antigen 9 Monoclonal specifically detects CD229/SLAMF3/Lymphocyte Antigen 9 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Especificaciones

Especificaciones

CD229/SLAMF3/Lymphocyte Antigen 9
Monoclonal
Alexa Fluor 647
50mM Sodium Borate
Rabbit
Protein A purified
RUO
4063
Store at 4°C in the dark.
IgG
Flow Cytometry, Immunofluorescence
2
Flow Cytometry, Immunocytochemistry/Immunofluorescence
CD229, CD229 antigen, cell-surface molecule Ly-9, hly9, lymphocyte antigen 9Cell surface molecule Ly-9, mLY9, SLAMF3, T-lymphocyte surface antigen Ly-9
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD229/SLAMF3/Lymphocyte Antigen 9 (NP_032560.2; Met1-Arg453).
0.1 mL
Primary
Mouse
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales