missing translation for 'onlineSavingsMsg'
Learn More

CD226 (DNAM-1) Mouse anti-Human, APC, Clone: 11A8.7.4, eBioscience™

Mouse Monoclonal Antibody

Marca:  Invitrogen 17-2269-42

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15913214

  • 230.00€ / paquete de 100


Fecha de envío estimada
del proveedor 06-05-2019
Añadir a la cesta

Descripción

Descripción

DNAM 1 is a receptor expressed by peripheral blood T lymphocytes that is involved in intercellular adhesion, lymphocyte signaling, cytotoxicity and lymphokine secretion mediated by cytotoxic T lymphocytes. DNAM 1 is broadly expressed on T cells, NK cells, platelets, monocytes and a subset of B cells. DNAM-1 is also expressed by a subset of CD3 positive thymocytes.
Especificación

Especificación

CD226 (DNAM-1)
Monoclonal
5 μL/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
Q15762
CD226
IgG1, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
10666
Flow Cytometry
11A8.7.4
APC
CD226
Liquid
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Videos