missing translation for 'onlineSavingsMsg'
Learn More

CD200R1 Antibody (OX108), Janelia Fluor™ 549 - Chimeric, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-52642JF549

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213371

  • 612.30€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

CD200R1 Monoclonal specifically detects CD200R1 in Human samples. It is validated for Flow Cytometry.
Especificaciones

Especificaciones

CD200R1
Monoclonal
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
CD200R1
Recombinant protein - hCD200RCD4d3+4.
0.1 mL
Primary
Recognizes human CD200R, a receptor for the CD200/OX2.
Store at 4°C in the dark.
IgG κ
Flow Cytometry
OX108
Flow Cytometry
CD200 receptor 1, CD200RMOX2Rcell surface glycoprotein CD200 receptor 1, Cell surface glycoprotein OX2 receptor 1, cell surface glycoprotein receptor CD200, CRTR2, HCRTR2, MOX2 receptor, OX2RCD200 cell surface glycoprotein receptor
Rabbit
Protein A or G purified
Cancer, GPCR, Neuronal Cell Markers, Neuroscience
131450
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales