Learn More
CD1d Monoclonal Antibody (1B1), Super Bright™ 645, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
en la oferta
Especificaciones
Antígeno | CD1d |
---|---|
Clon | 1B1 |
Concentración | 0.2 mg/mL |
Aplicaciones | Flow Cytometry |
Clasificación | Monoclonal |
Descripción
Description: This L161 monoclonal antibody detects CD1c (also known as BDCA-1), a glycoprotein that is noncovalently linked to beta-2 microglobulin on thymocytes and antigen presenting cells such as dendritic and Langerhans cells. This molecule is also expressed on some circulating and marginal zone B cells, as well as in lymph nodes and germinal centers. CD1c is involved in the presentation of lipid antigens such as microbial fatty acids to effector T cells during the adaptive immune response. Finally, alternative splicing gives rise to three different isoforms of CD1c (soluble, membrane, and cytoplasmic/soluble isoforms). Applications Reported: This L161 antibody has been reported for use in flow cytometric analysis. Applications Tested: This L161 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 645 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 645 nm. We recommend using a 660/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome.
CD1d Monoclonal antibody specifically detects CD1d in Mouse samples. It is validated for Flow Cytometry.Especificaciones
CD1d | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
P11609 | |
Cd1d1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Cd1d1 |
1B1 | |
Flow Cytometry | |
Super Bright 645 | |
Rat | |
Mouse | |
12479 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |