missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Rat, anti-Mouse, Alexa Fluor 488, Clone: 6D5, STEMCELL Technologies
Rat Monoclonal Antibody
182.25€ - 440.10€
Especificaciones
Antígeno | CD19 |
---|---|
Clon | 6D5 |
Aplicaciones | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
Clasificación | Monoclonal |
Conjugado | Alexa Fluor 488 |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
17108371
|
Stemcell Technologies
60006AD.1 |
25 μg |
182.25€
25 microgramos |
Fecha estimada de envío: 09-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
17198361
|
Stemcell Technologies
60006AD |
100 μg |
440.10€
100 microgramos |
Fecha estimada de envío: 09-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
- The 6D5 antibody reacts with CD19, an ∼95 kDa type 1 transmembrane glycoprotein expressed on the surface of B cells throughout all stages of development, from early pre-B cells to plasma cells
- Expression is down-regulated but persists in terminally differentiated plasma cells
- CD19 is also found on follicular dendritic cells
- By associating with CD21 and CD81, CD19 functions as a co-receptor for the B cell receptor and is involved in B cell activation and differentiation
- Activation of CD19 is accompanied by phosphorylation of the cytoplasmic domain, which promotes binding to kinases and the induction of intracellular signaling cascades
- Mutations in CD19 can result in severe immunodeficiency syndromes
- Clone 6D5 recognizes the same epitope as clone ID3 in cross-competition assays
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse B Cell Isolation Kit (Catalog #19854)
Especificaciones
CD19 | |
Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
Alexa Fluor 488 | |
Mouse | |
Primary |
6D5 | |
Monoclonal | |
Rat | |
IgG2a κ |