Learn More
CD186 (CXCR6) Rat anti-Mouse, PE, Clone: DANID2, eBioscience™
Rat Monoclonal Antibody
118.00€ - 279.00€
Productos 2
Código de producto Marca Cantidad. Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad. Precio Cantidad y disponibilidad  
15933184 Ver documentos Invitrogen
12-9186-80
25 μg 118.00€
25 microgramos Ver productos alternativos
Ahorre hasta 
»
Fecha de envío estimada
del proveedor 28-10-2019
Añadir a la cesta
15943184 Ver documentos Invitrogen
12-9186-82
100 μg 279.00€
100 microgramos Ver productos alternativos
Ahorre hasta 
»
Fecha de envío estimada
del proveedor 28-10-2019
Añadir a la cesta
Descripción
Descripción
CD186, also known as CXCR6, is a G protein-coupled chemokine receptor reported to be one of the cellular receptors of HIV and SIV. CD186 is expressed by NKT cells, various subtypes of T cells with activated and memory phenotypes, and some plasma cells. It has been reported that mouse liver NK cells, but not naïve or splenic NK cells, express CD186. The ligand of CD186, CXCL16, is expressed by macrophages, dendritic cells and liver endothelial cells. Being highly expressed on NKT cells, CD186 is often used as a proxy marker of this cell subset.