missing translation for 'onlineSavingsMsg'
Learn More

CD186 (CXCR6) Rat anti-Mouse, APC, Clone: DANID2, eBioscience™

Rat Monoclonal Antibody

Marca:  Invitrogen 17-9186-82

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15913224

  • 287.00€ / 100 microgramos


Fecha de envío estimada
del proveedor 03-05-2019
Añadir a la cesta

Descripción

Descripción

CD186, also known as CXCR6, is a G protein-coupled chemokine receptor reported to be one of the cellular receptors of HIV and SIV. CD186 is expressed by NKT cells, various subtypes of T cells with activated and memory phenotypes, and some plasma cells. It has been reported that mouse liver NK cells, but not naïve or splenic NK cells, express CD186. The ligand of CD186, CXCL16, is expressed by macrophages, dendritic cells and liver endothelial cells. Being highly expressed on NKT cells, CD186 is often used as a proxy marker of this cell subset.
Especificación

Especificación

CD186 (CXCR6)
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q9EQ16
Bonzo, STRL33, TYMSTR
Rat
IgG2a, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
80901
Flow Cytometry
DANID2
APC
CXCR6
Liquid
CXCR6
Mammalian cell line expressing the full form of CD186
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Videos