Learn More
CD178 / Fas ligand Mouse, APC, Clone: NOK-1, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio QAB85-APC-100Tests
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CD178 / Fas ligand | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
FASLG | |
Protein A/G Purified | |
RUO | |
356 | |
IgG1 κ |
Flow Cytometry | |
NOK-1 | |
APC | |
NP_001289675 | |
Mouse | |
100 Tests | |
Primary | |
Store at 2-4°C. |