Learn More
CD16 Monoclonal Antibody (3G8), Super Bright™ 600, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
en la oferta
Especificaciones
Antígeno | CD16 |
---|---|
Clon | 3G8 |
Concentración | 5 μL/Test |
Aplicaciones | Flow Cytometry |
Clasificación | Monoclonal |
Descripción
Description: This 3G8 monoclonal antibody reacts with human and non-human primate CD16, which is also known as the low-affinity Fc gamma RIII. CD16 exists as two distinct isoforms, Fc gamma RIIIA and Fc gamma RIIIB. In humans, Fc gamma RIIIA is expressed as a polypeptide-anchored form on monocytes, macrophages, and lymphocytes such as NK cells. T and B cells do not express this Fc receptor. Fc gamma RIIIB is also detected on neutrophils as a GPI-anchored form. Expression of CD16 on lymphocytes and monocytes is similar in non-human primates. However, while CD16 is not found on neutrophils in macaques and baboons, this receptor is detected on these cells in sooty mangabeys. Binding of IgG leads to activation of signal transduction pathways, resulting in antibody-dependent cell-mediated cytotoxicity (ADCC), phagocytosis, cytokine release, and antigen presentation. This monoclonal antibody has been reported to have several functional activities, including inhibition of cytotoxic ability, activation of cell signaling, and NK cell depletion in vivo. Moreover, the 3G8 antibody clone has been demonstrated to work on capuchin monkey, chimpanzee, common marmoset, cynomologous monkey, hamadyras baboon, olive baboon, pigtailed macaque, rhesus, and squirrel monkey. Based on cross-blocking studies 3G8 recognizes the same epitope as CB16. However, 3G8 and B73.1 antibody clones bind distinct epitopes.
CD16 Monoclonal antibody specifically detects CD16 in Baboon, Chimpanzee, Cynomolgus Monkey, Human, Non-human Primate, Rhesus Monkey samples. It is validated for Flow Cytometry.Especificaciones
CD16 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
IgG1 | |
Affinity chromatography | |
Antibody |
3G8 | |
Flow Cytometry | |
Super Bright 600 | |
Mouse | |
Baboon, Chimpanzee, Cynomolgus Monkey, Human, Non-human Primate, Rhesus Monkey | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
FCGR3B |