Learn More
CD14 Mouse anti-Human, Super Bright 780, Clone: 61D3, eBioscience™
Mouse Monoclonal Antibody
145.00€ - 282.00€
Productos 2
Código de producto Marca Cantidad. Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad. Precio Cantidad y disponibilidad  
15934134 Ver documentos Invitrogen
78-0149-41
25 Tests 145.00€
paquete de 25 Ver productos alternativos
Ahorre hasta 
»
Fecha de envío estimada
del proveedor 10-06-2020
Añadir a la cesta
15944134 Ver documentos Invitrogen
78-0149-42
100 Tests 282.00€
paquete de 100 Ver productos alternativos
Ahorre hasta 
»
Fecha de envío estimada
del proveedor 10-06-2020
Añadir a la cesta
Descripción
Descripción
In some experiments, we have observed that compensation values for Super Bright 780-conjugated antibodies are higher in the violet 450/50 channel when using UltraComp eBeads microspheres (Product No. 01-2222-42) as compared to single-color stained cells. In such circumstances, we would recommend setting compensation with cells. We have also observed this in some experiments using AbC Total Antibody Compensation beads (Product No. A10497). Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 780 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company. CD14 is a 55 kDa GPI-anchored glycoprotein that is constitutively expressed on the surface of mature monocytes, macrophages, and neutrophils. CD14 also serves as a multifunctional lipopolysaccharide receptor, and is released to the serum both as a secreted and enzymatically cleaved GPI-anchored form. CD14 binds lipopolysaccharide molecule in a reaction catalyzed by lipopolysaccharide-binding protein (LBP), an acute phase serum protein. The soluble sCD14 can discriminate slight structural differences between lipopolysaccharides and is important for neutralization of serum allochthonous lipopolysaccharides by reconstituted lipoprotein particles. Further, CD14 has been shown to bind apoptotic cells, and can affect allergic, inflammatory and infectious processes. Alternative splicing results in multiple transcript variants encoding the same CD14 isoform. Diseases associated with CD14 dysfunction include mycobacterium chelonae infection and Croup.