missing translation for 'onlineSavingsMsg'
Learn More

CD137 (4-1BB), Super Bright 600, Clone: 4B4 (4B4-1), Monoclonal, eBioscience™

Mouse Monoclonal Antibody

Marca:  Invitrogen 63-1379-41

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923534

  • 147.00€ / paquete de 25


Fecha de envío estimada
del proveedor 02-04-2019
Añadir a la cesta

Descripción

Descripción

We recommend using a 610/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No. SB-4400) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 600 nm; Laser: Violet Laser The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor contributes to the clonal expansion, survival, and development of T cells. It can also induce proliferation in peripheral monocytes, enhance T cell apoptosis induced by TCR/CD3 triggered activation, and regulate CD28 co-stimulation to promote Th1 cell responses. The expression of this receptor is induced by lymphocyte activation. TRAF adaptor proteins have been shown to bind to this receptor and transduce the signals leading to activation of NF-kappaB.
Especificación

Especificación

CD137 (4-1BB)
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
Q07011
TNFRSF9
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3604
Flow Cytometry
4B4 (4B4-1)
Super Bright 600
TNFRSF9
Liquid
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Videos