Learn More
CD11c Hamster, anti-Mouse, PerCP-Cyanine5.5, Clone: N418, STEMCELL Technologies
Hamster Monoclonal Antibody
Marca: Stemcell Technologies 60002PS.1
Detalles adicionales : Peso : 0.00110kg
Descripción
- The N418 antibody reacts with CD11c (αX integrin), a 150 kDa type 1 transmembrane glycoprotein that associates non-covalently with CD18 (β2 integrin) to form a heterodimeric cell surface adhesion receptor
- Through its interaction with ligands such as iC3b, fibrinogen, and CD54, the CD11c/CD18 receptor is involved in several immune response processes, including cell migration, stimulation of cytokine production by monocytes and macrophages, T cell proliferation, leukocyte recruitment, and phagocytosis
- In mice, CD11c is expressed on dendritic cells, macrophages, monocytes, granulocytes, NK cells, and a subset of T cells
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse CD11c Positive Selection Kit II (Catalog #18780)
Especificaciones
CD11c | |
Monoclonal | |
PerCP-Cyanine5.5 | |
25 μg | |
Mouse | |
Dendritic Cells |
CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
N418 | |
Hamster | |
Primary | |
IgG |