Learn More
CD11b Mouse, PE-Cy7, Clone: ICRF44, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio QAB25-PE7-25Tests
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CD11b | |
ICRF44 | |
PE-Cyanine7 | |
XP_006721108 | |
Mouse | |
Protein A/G Purified | |
RUO | |
Primary | |
3684 |
Flow Cytometry | |
5μL/Test | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
ITGAM | |
IgG1 kappa | |
25 Tests | |
Store at 2-4°C. | |
Monoclonal |