missing translation for 'onlineSavingsMsg'
Learn More

CD117 (c-Kit), Super Bright 780, Clone: 2B8, Monoclonal, eBioscience™

Rat Monoclonal Antibody

Marca:  Invitrogen 78-1171-80

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15914164

  • 135.00€ / 25 microgramos


Fecha de envío estimada
del proveedor 01-04-2019
Añadir a la cesta

Descripción

Descripción

When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No. SB-4400) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. In some experiments, we have observed that compensation values for Super Bright 780-conjugated antibodies are higher in the violet 450/50 channel when using UltraComp eBeads microspheres (Product No. 01-2222-42) as compared to single-color stained cells. In such circumstances, we would recommend setting compensation with cells. We have also observed this in some experiments using AbC Total Antibody Compensation beads (Product No. A10497). Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 780 nm; Laser: Violet Laser c-Kit, also known as CD117 and stem cell factor receptor, is a 145 kDa transmembrane tyrosine kinase encoded by the c-Kit proto-oncogene. c-Kit acts to regulate a variety of biological responses including cell proliferation, apoptosis, chemotaxis and adhesion. Ligand binding to the extracellular domain leads to autophosphorylation on several tyrosine residues within the cytoplasmic domain, and activation. c-Kit mutations correlate with tumor growth and progression in a variety of cancers including mast cell disease, gastrointestinal stromal tumor, acute myeloid leukemia, Ewing sarcoma, and lung cancer. Phosphorylation at tyrosine 703 of c-Kit allows binding of Grb2 and activation of the Ras-Raf-ERK1 and 2 signaling pathway.
Especificación

Especificación

CD117 (c-Kit)
Monoclonal
0.2 mg/mL
PBS with BSA and 0.09% sodium azide; pH 7.2
P05532, Q2HWD6
C-Kit, CD117, PBT, SCFR, mast/stem cell growth factor receptor Kit, p145 c-kit, piebald trait protein, proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, soluble KIT variant 1, tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein, ckit
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse, Porcine
Flow Cytometry
2B8
Super Bright 780
Kit
Liquid
KIT
IgG2b, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
16590, 396810
Videos