Learn More
CD117/c-kit Antibody (YB5.B8), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Novus Biologicals NBP1-43358UV
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18213283
  • 562.62€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock
Descripción
Descripción
CD117/c-kit Monoclonal specifically detects CD117/c-kit in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Especificaciones
Especificaciones
CD117/c-kit
Monoclonal
DyLight 350
CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein
Mouse
Affinity Purified
RUO
Primary
Human
IgG1 κ
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
YB5.B8
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
KIT
Human CD117
0.1 mL
Cancer, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers, Tyrosine Kinases
3815
Store at 4C in the dark.