missing translation for 'onlineSavingsMsg'
Learn More

CD105 (Endoglin), Super Bright 645, Clone: SN6, Monoclonal, eBioscience™

Mouse Monoclonal Antibody

Marca:  Invitrogen 64-1057-41

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923634

  • 138.00€ / paquete de 25


Fecha de envío estimada
del proveedor 03-04-2019
Añadir a la cesta

Descripción

Descripción

Light sensitivity: This tandem dye is sensitive to photo- induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 645 nm; Laser: Violet Laser CD105 or Endoglin is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists on an O- and N-glycosylated homodimer. Up regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation associated endothelial marker. CD105 binds to TGF beta 1 and 3 with high affinity but not to TGF beta 2.
Especificación

Especificación

CD105 (Endoglin)
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
P17813
ENG
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
2022
Flow Cytometry
SN6
Super Bright 645
ENG
Liquid
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Videos