missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Ccl3 (Mouse) ELISA Kit

In-Vitro Enzyme-Linked Immunosorbent assay for the quantitative measurement of Mouse Ccl13

Marca:  Abnova™ KA1804.1Kit

Detalles adicionales : Peso : 0.02000kg

 Ver más versiones de este producto

Código de producto. 16056835

  • 578.00€ / 1 set


Fecha estimada de envío
19-09-2019
Añadir a la cesta

Especificaciones

Especificaciones

Biotin
Ccl3 (Mouse) ELISA Kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse Ccl3.
Colorimetric
96-well Antibody coated microwell, 25mL Wash Buffer 20X, 2 vials standard, 30mL assay diluent A, 15mL assay diluent B,, 2 vials detection Antibody MSP,8μL HRP Streptavidian concentrrate,12mL TMB one step Substrate reagent,8mL stop soluti
96 Assays
Store the kit at -20°C. Standard should be stored at -20°C or -80°C. Avoid repeated freezing and thawing.
Mouse
8pg/mL
Quantification
Ccl3 (Mouse) ELISA Kit
Cell Culture Supernatant, Plasma, Serum
100μL
Videos
Documentos