missing translation for 'onlineSavingsMsg'
Learn More

CCL28 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 18214-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16818274

  • 109.00€ / 20 microlitros

Envío estimado: 22-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene belongs to the subfamily of small cytokine CC genes. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity for resting CD4 or CD8 T cells and eosinophils. The product of this gene binds to chemokine receptors CCR3 and CCR10. This chemokine may play a role in the physiology of extracutaneous epithelial tissues, including diverse mucosal organs.
Especificaciones

Especificaciones

CCL28
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9JIL2, Q9NRJ3
C C motif chemokine 28, CCK1, CCL28, MEC, Protein CCK1, SCYA28, Small inducible cytokine A28
Rabbit
IgG
20 μL
-20°C
Primary
56477, 56838
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CCL28
Liquid
CCL28
CCL28 Fusion Protein Ag12920
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Human
Videos
SDS
Documentos
Certificados