missing translation for 'onlineSavingsMsg'
Learn More

CCDC47 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NBP2-82078

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18216299

  • 469.75€ / 0.10 miligramo
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

CCDC47 Polyclonal specifically detects CCDC47 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

CCDC47
Polyclonal
Unconjugated
PBS
CCDC47
CCDC47 antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human CCDC47. The immunogen is located within amino acids 350 - 400 of CCDC47. Amino Acid Squence: NVPGSGNTYPKDMEAL
0.1 mg
Primary
Human, Mouse
IgG
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
1 mg/mL
Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin
coiled-coil domain containing 47, coiled-coil domain-containing protein 47, GK001, MSTP041, PSEC0077
Rabbit
Affinity Purified
RUO
57003
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales