Learn More
CCDC43 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca:  Bio-Techne NB100-97844
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18214410
  • 534.54€ / 100 microlitros
Fecha estimada de envío: 14-08-2024
para ver el stock
Descripción
Descripción
CCDC43 Polyclonal specifically detects CCDC43 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Especificaciones
Especificaciones
CCDC43
Polyclonal
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
coiled-coil domain containing 43, coiled-coil domain-containing protein 43, FLJ31795
Rabbit
Affinity Purified
RUO
124808
Store at 4C. Do not freeze.
Western Blot, Immunoprecipitation
1.0 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation
NP_653210.2
CCDC43
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human coiled-coil domain-containing protein 43 using the numbering given in entry NP_653210.2 (GeneID 124808).