missing translation for 'onlineSavingsMsg'
Learn More

CBR1 Antibody (4E12), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NBP1-74041AF700

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213585

  • 521.58€ / 100 microlitros
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

CBR1 Monoclonal specifically detects CBR1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Especificaciones

Especificaciones

CBR1
Monoclonal
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
CBR1
Recombinant human CBR1 (1-277 aa ) purified from E. coli
100 μL
Stem Cell Markers
873
Human, Mouse
Purified
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
4000000000000
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
15-hydroxyprostaglandin dehydrogenase [NADP+], carbonyl reductase (NADPH) 1, carbonyl reductase (NADPH) 1, EC 1.1.1.18410EC 1.1.1.197,15-hydroxyprostaglandin dehydrogenase, carbonyl reductase [NADPH] 1, carbonyl reductase 1, CBR, CRN, EC 1.1.1.184, EC 1.1.1.189, hCBR1, NADPH-dependent carbonyl reductase 1, Prostaglandin 9-ketoreductase, Prostaglandin-E(2) 9-reductase, SDR21C1, short chain dehydrogenase/reductase family 21C, member 1
Mouse
Protein G purified
RUO
Primary
Anti-human CBR1 mAb, clone 4E12, is derived from hybridization of mouse SP2/0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human CBR1 protein.
Store at 4C in the dark.
IgG2a κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales