missing translation for 'onlineSavingsMsg'
Learn More

Cathepsin K Antibody (CTSK/2791), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP2-79889B

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215747

  • 578.82€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

Cathepsin K Monoclonal specifically detects Cathepsin K in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

Cathepsin K
Monoclonal
Biotin
PBS with 0.05% Sodium Azide
CTSK
Recombinant fragment of human Cathepsin K protein (around aa 163-274) (exact sequence is proprietary) (Uniprot: P43235)
0.1 mL
Cancer
Human
IgG1 κ
Immunohistochemistry, Immunohistochemistry (Paraffin)
CTSK/2791
Immunohistochemistry, Immunohistochemistry-Paraffin
cathepsin K, cathepsin K (pycnodysostosis), Cathepsin O, cathepsin O1, Cathepsin O2, Cathepsin X, CTS02, CTSO, CTSO1, CTSO2, EC 3.4.22, EC 3.4.22.38, MGC23107, PKND, PYCD
Mouse
Protein A or G purified
RUO
Primary
Store at 4°C in the dark.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales