missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Catenin, beta (CTNNB1) Mouse, Clone: CTNNB1/2098, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 1499-MSM11-P0
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Catenin, beta (CTNNB1) | |
Cell surface and cytoplasmic | |
CTNNB1/2098 | |
Unconjugated | |
Mouse | |
92kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
1499 | |
IgG2b κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant full-length human beta-Catenin (CTNNB1) protein | |
HeLa or MCF-7 cells. Breast carcinoma | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |