missing translation for 'onlineSavingsMsg'
Learn More

Casein Kinase 1 alpha Antibody (2E2), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP1-30165JF549

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213525

  • 521.58€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Description

Description

Casein Kinase 1 alpha Monoclonal specifically detects Casein Kinase 1 alpha in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

CaseinKinase1alpha
Monoclonal
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
CSNK1A1
Recombinant human Casein Kinase 1 alpha (1-337aa) purified from E. coli
0.1 mL
Primary
This antibody is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human Casein Kinase 1 alpha protein.
Store at 4°C in the dark.
IgG2b κ
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
200
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
casein kinase 1, alpha 1, casein kinase I isoform alpha, CK1CKI-alpha, down-regulated in lung cancer, EC 2.7.11.1, HLCDGP1, PRO2975
Mouse
Protein G purified
Protein Kinase, Wnt Signaling Pathway
1452
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers