Learn More
Casein, Hammarsten Grade, Thermo Scientific Chemicals
Cream to light yellow, lyophilized powder.
Marca: Thermo Scientific J12840.TD
Detalles adicionales : CAS : 9000-71-9 Peso : 1.00000kg
Descripción
- Cream to light yellow
- Protein: ≥95%
- Melting point: ∽280°C (dec.)
- MDLNumber: MFCD00081481
- CAS: 9000-71-9
Especificaciones
75000 to 100000 | |
Plastic Bottle | |
Ambient Temperatures, Store under Nitrogen | |
Lyophilized |
1 lb. | |
Casein | |
Hygroscopic |
RUO – Research Use Only