missing translation for 'onlineSavingsMsg'
Learn More

CARD6 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 18029-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16818064

  • 109.00€ / 20 microlitros

Envío estimado: 27-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a protein that contains a caspase recruitment domain (CARD), an antiparallel six-helical bundle that mediates homotypic protein-protein interactions. The encoded protein is a microtubule-associated protein that has been shown to interact with receptor-interacting protein kinases and positively modulate signal transduction pathways converging on activation of the inducible transcription factor NF-kB.
Especificaciones

Especificaciones

CARD6
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BX69
CARD6, CINCIN1
Rabbit
IgG
20 μL
-20°C
Primary
84674
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
CARD6
Liquid
CARD6
CARD6 Fusion Protein Ag12595
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documentos
Certificados