Learn More
Swann-Morton™ Carbon Steel Postmortem Scalpel Blades
Product Code. 12952555
Change view
Click to view available options
Size:
# PM40
# PM40 B
Product Type:
Blunted Knife
Sharp
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Product Type Unit Size
12952555 # PM40 B Blunted Knife Pack of 10
11360905 # PM40 Sharp Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12952555 Supplier Swann-Morton™ Supplier No. 2552
  • ONLINE EXCLUSIVE PRICE
    17.40€
    15.49€ / paquete de 10
    Ahorro 1.91€
    10% Descuento
View all promotions
Fecha estimada de envoi: 18-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Use Swann Morton™ Carbon Steel Postmortem Scalpel Blades to facilitate pathology operations.
  • Added safety feature of a 'bull nose tip' to help offset the occurrence of accidental sharp related injuries
  • Blunted
  • PM-40
Specifications
Blade Material Carbon Steel
Product Type Blunted Knife
For Use With (Application) For Postmortem procedures with the added safety feature of a 'bull nose tip' to help offset the occurrence of Accidental Sharp related Injuries
Size # PM40 B
For Use With (Equipment) The PM40 Stainless, PM40 Standard or PM40 Plated Handle