Learn More
Carbon Dioxide Standard (CO2) Release Agent, Reagecon
BIOPHARMA
Product Code. 11513052 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
6 x 200 mL
Unit Size:
Pack of 6
Product Code. Quantity Unit Size
11513052 6 x 200 mL Pack of 6
1 options
This item is not returnable. View return policy
Product Code. 11513052 Supplier Reagecon Supplier No. RR021
  • ONLINE EXCLUSIVE PRICE
    216.00€
    162.19€ / paquete de 6
    Ahorro 53.81€
    24% Descuento
 Restricted Item
Fecha estimada de envoi: 02-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Used as an acidic CO2 release agent during the measurement of CO2 using the Corning 965D Carbon Dioxide Analyzer
  • Use of this instrument is one of a number of approved techniques available for the determination of carbon dioxide, in beer and other carbonated beverages.
  • This instrument was subsequently marketed by Mettler Toledo as the model M965D CO2 Analyzer.
  • Produced using raw materials of a very high purity and specification.
  • Tested using Reagecon Test Method TPATA with results expressed in molar acidity, specification 2.0 – 2.2 M acidity.
  • A Certificate of Conformity is issued with each batch.
Specifications
For Use With (Application) Carbon Dioxide Analysis
Quantity 6 x 200 mL
compliance-icons
Product Identifier
  • CO2 RELEASE REAGENT
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Supplemental information
  • MIXTURE LIST-Contain: Lactic acid