Learn More
Éster de pinacol de ácido carbazol-2-borónico, téc. 90 %, Thermo Scientific™
Catalog Number p-7052718
Click to view available options
Cantidad:
250 mg
1 g
871125-67-6
C18H20BNO2
293.173
MFCD07784368
RLSJGSFDSSYNPL-UHFFFAOYSA-N
9h-carbazole-2-boronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9h-carbazole, 9h-carbazole,4a,9a-dihydro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, carbazole-2-boronic acid pinacol ester, 9h-carbazol-2-ylboronic acid pinacol ester, 9h-carbazol-2-yl boronic acid pinacol ester, 2-tetramethyl-1,3,2-dioxaborolan-2-yl-9h-carbazole, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-9h-carbazole, 2-9h-carbazol-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
46738013
2-(4,4,5,5-tetrametil-1,3,2-dioxaborolan-2-il)-9H-carbazol
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4N3
871125-67-6
C18H20BNO2
293.173
MFCD07784368
RLSJGSFDSSYNPL-UHFFFAOYSA-N
9h-carbazole-2-boronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9h-carbazole, 9h-carbazole,4a,9a-dihydro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, carbazole-2-boronic acid pinacol ester, 9h-carbazol-2-ylboronic acid pinacol ester, 9h-carbazol-2-yl boronic acid pinacol ester, 2-tetramethyl-1,3,2-dioxaborolan-2-yl-9h-carbazole, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-9h-carbazole, 2-9h-carbazol-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
46738013
2-(4,4,5,5-tetrametil-1,3,2-dioxaborolan-2-il)-9H-carbazol
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4N3
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Carbazole-2-boronic acid pinacol ester
Cantidad 250 mg
Formula Weight (peso de la fórmula) 293.17
Porcentaje de pureza 90%
Grado Técnico
Forma física Sólido