missing translation for 'onlineSavingsMsg'
Learn More

Erlab™ Captair™ Smart 481 Ductless Fume Hood

Designed with advanced carbon filtration to minimize chemical exposure.

Marca:  Erlab™ A62A/BE

Detalles adicionales : Peso : 90.00000kg

 Ver más versiones de este producto

Código de producto. 15564360

  • 4981.20€ / cada uno


Consúltenos : 902 239 303 
Añadir a la cesta

Descripción

Descripción

  • Sensors to detect filter breakthrough of solvents, acids or formaldehyde
  • Safety back up filter in case of main filter saturation
  • Continuous monitoring of air face velocity

Erlab Safety Program

  • Application analysis and validation, usage certification, filter change reminders
  • Connect with eGuard™ for real-time status, notifications and alerts
  • Smart-Light feature for lighting up the hood

Flexibility

  • Configurable filtration column
  • No ductwork needed


Environment

  • No dangerous chemicals are released into atmosphere
  • Low energy consumption
Especificaciones

Especificaciones

135 cfm
53.3 cm
Class2, type 1C, carbon filters, liquid chemical handling
128.5 cm
Two carbon filter
117.6 cm
61.5 cm
220V 50/60Hz, 65w
For organic vapor, acid
110.0. cm
124.5 cm
Ductless filter hood
Videos
Documentos