Learn More
Erlab™ Captair™ Pyramid PVC Glove Bag for Fume Hood
Marca: Erlab™ 2200ANM
Detalles adicionales : Peso : 3.17515kg
Especificaciones
72.39 cm | |
55.88 cm | |
Pyramid™ Glove Bag | |
Ideal for research performed on-location and in the laboratory and for protection of operators, protection of samples |
Butyl Gloves, 0.12 in. I.D. Coupler Body Valved Fitting, Flexible PVC Protective Package | |
76.2 cm | |
PVC |