Learn More
Cantharidin, Mikromol™
Description
Cantharidin
Specifications
Specifications
| Chemical Name or Material | Cantharidin |
| CAS | 56-25-7 |
| Analyte or Component Names | Cantharidin |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H12 O4 |
| Certifications/Compliance | ISO 17034 |
| Synonym | 7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic anhydride, 2,3-dimethyl-, Cantharidin, 1,2-Dimethyl-3,6-epoxyperhydrophthalic anhydride, Canthacur, Cantharides camphor, Cantharidine, Cantharone, Hexahydro-3a,7a-dimethyl-4,7-epoxyisobenzofuran-1,3-dione, Kantaridin, NSC 61805, exo-1,2-cis-Dimethyl-3,6-epoxyhexahydrophthalic anhydride, 4,7-Epoxyisobenzofuran-1,3-dione, hexahydro-3a,7a-dimethyl-, (3aalpha,4beta,7beta,7aalpha)- |
| InChI Formula | InChI=1S/C10H12O4/c1-9-5-3-4-6(13-5)10(9,2)8(12)14-7(9)11/h5-6H,3-4H2,1-2H3/t5-,6+,9+,10- |
| SMILES | C[C@@]12[C@H]3CC[C@H](O3)[C@]1(C)C(=O)OC2=O |
| IUPAC Name | (3aα,4β,7β,7aα)-Hexahydro-3a,7a-dimethyl-4,7-epoxyisobenzofuran-1,3-dione |
| Show More |