missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CAMK2A (Human) Recombinant Protein
Human CAMK2A full-length ORF ( AAH40457.1, 1 a.a. - 478 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00000815-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MATITCTRFTEEYQLFEELGKGAFSVVRRCVKVLAGQEYAAKIINTKKLSARDHQKLEREARICRLLKHPNIVRLHDSISEEGHHYLIFDLVTGGELFEDIVAREYYSEADASHCIQQILEAVLHCHQMGVVHRDLKPENLLLASKLKGAAVKLADFGLAIEVEGEQQAWFGFAGTPGYLSPEVLRKDPYGKPVDLWACGVILYILLVGYPPFWDEDQHRLYKQIKAGAYDFPSPEWDTVTPEAKDLINKMLTINPSKRITAAEALKHPWISHRSTVASCMHRQETVDCLKKFNARRKLKGAILTTMLATRNFSGGKSGGNKKSDGVKESSESTNTTIEDEDTKVRKQEIIKVTEQLIEAISNGDFESYTKMCDPGMTAFEPEALGNLVEGLDFHRFYFENLWSRNSKPVHTTILNPHIHLMGDESACIAYIRITQYLDAGGIPRTAQSEETRVWHRRDGKWQIVHFHRSGAPSVLPH
Especificaciones
AAH40457.1 | |
calcium/calmodulin-dependent protein kinase II alpha | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAMK2A | |
GST |
815 | |
Wheat germ expression system | |
25 μg | |
CAMKA, KIAA0968 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |