missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Human CAMK2A (CaMKII Alpha), His Tag Recombinant Protein

Human CAMK2A (CaMKII Alpha), His Tag Recombinant Protein

Marca:  Thermo Scientific™ PV3142

Detalles adicionales : Peso : 3.36000kg

 Ver más versiones de este producto

Código de producto. 11539326

  • 638.00€ / 10 microgramos
En stock
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Autophosphorylation of threonine 286 in the presence of Ca2+ and calmodulin activates CAMK2A with the ability to switch from calmodulin-dependent to a calmodulin independent activity.
Especificaciones

Especificaciones

NP_741960
25400, 815, 816, 817, 818
10 μg
CAMK2 Kinase Family
Recombinant
Storage Buffer: 20 mM Tris pH 7.5, 100 mM NaCl, 0.5 mM EDTA, 0.05% NP-40, 2 mM DTT and 50% Glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Ser⁄Thr Kinases: CAMK Group
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate
calcium⁄calmodulin-dependent protein kinase (CaM kinase) II alpha
Kinases & Inhibitors
Liquid
Signal Transduction
Kinase Assay
56.8 kDa
CAMK2A (CaMKII Alpha)
Human
His Tag
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
Dry Ice
CAMK2A
Baculovirus
Recombinant, Full Length
Serine/Threonine Kinases
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales