missing translation for 'onlineSavingsMsg'
Learn More

Calpain 3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 10415-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16816793

  • 319.00€ / 150 microlitros

Envío estimado: 28-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Calpain, a heterodimer consisting of a large and a small subunit, is a major intracellular protease, although its function has not been well established. This gene encodes a muscle-specific member of the calpain large subunit family that specifically binds to titin. Mutations in this gene are associated with limb-girdle muscular dystrophies type 2A. Alternate promoters and alternative splicing result in multiple transcript variants encoding different isoforms and some variants are ubiquitously expressed.
Especificaciones

Especificaciones

Calpain 3
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P16259, P20807, Q64691
Calpain 3, calpain 3, (p94), Calpain L3, Calpain p94, CANP 3, CANP3, CANPL3, CAPN3, LGMD2, LGMD2A, nCL 1, NCL1, New calpain 1, p94
Rabbit
IgG
150 μL
-20°C
Primary
12335, 29155, 825
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CAPN3
Liquid
Capn3
Calpain 3 Fusion Protein Ag0671
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documentos
Certificados