Learn More
Hach Lange™ Calibration Set
Product Code. 10587354
Change view
Click to view available options
Volume (Metric):
100 mL
Unit Size:
Each
Product Code. Volume (Metric) Unit Size
10587354 100 mL Each
1 options
This item is not returnable. View return policy
Product Code. 10587354 Supplier Hach Lange™ Supplier No. 2659410
Fecha estimada de envoi: 21-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Hach Lange Calibration Set, Volume: 100mL
TRUSTED_SUSTAINABILITY
Specifications
Type Calibration Kit
For Use With (Equipment) Hach Turbidimeters
Volume (Metric) 100 mL