missing translation for 'onlineSavingsMsg'
Learn More

Calcium Nitrate Tetrahydrate, 99.0 to 103.0%, ACS Reagent, Honeywell Fluka™

Catalog Number 15612360
Click to view available options
Cantidad:
500 g
2.5 kg
Envase:
Poly Bottle
Este artículo no se puede devolver. Vea la política de devoluciones
13477-34-4
CaH8N2O10
236.146
MFCD00149604
ICSSIKVYVJQJND-UHFFFAOYSA-N
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
16211656
CHEBI:86159
calcium;dinitrate;tetrahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2]
Este artículo no se puede devolver. Vea la política de devoluciones
13477-34-4
CaH8N2O10
236.146
MFCD00149604
ICSSIKVYVJQJND-UHFFFAOYSA-N
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
16211656
CHEBI:86159
calcium;dinitrate;tetrahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2]

ACS Reagent, 99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Calcium Nitrate Tetrahydrate
Punto de fusión 44°C
pH 4 to 7 at 20°C
Cantidad 2.5 kg
Fórmula lineal Ca(NO3)2 · 4H2O
Número UN NONH for all modes of transport
Formula Weight (peso de la fórmula) 236.15g/mol
Porcentaje de pureza 99.0 to 103.0%
Grado ACS Reagent
Envase Poly Bottle
Densidad 1.86g/mL at 25°C
Mostrar más Mostrar menos
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'